Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POU5F1B Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524309

Catalog No. PIPA524309

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine, mouse and porcine based on sequence homology.

This intronless gene was thought to be a transcribed pseudogene of POU class 5 homeobox 1, however, it has been reported that this gene can encode a functional protein. The encoded protein is nearly the same length as and highly similar to the POU class 5 homeobox 1 transcription factor, has been shown to be a weak transcriptional activator and may play a role in carcinogenesis and eye development.
Specifications

Specifications

POU5F1B
Polyclonal
PBS with 0.09% sodium azide
Q06416
OCT4-PG1, OTF3C, OTF3P1, POU5F1P1, POU5F1P4, POU5FLC20, POU5FLC8, POU 5 domain protein, POU class 5 homeobox 1 pseudogene 1, POU domain transcription factor OCT4-pg1, POU domain transcription factor Oct-4, POU domain, class 5, transcription factor 1 pseudogene 1, octamer binding protein 3_like sequence, putative POU domain, c
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5462
Flow Cytometry, Western Blot
Unconjugated
POU5F1B
Liquid
POU5F1B
KLH conjugated synthetic peptide between 214-242 amino acids from the C-terminal region of human P5F1B
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit