Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p62/SQSTM1 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP142822B
Description
p62/SQSTM1 Polyclonal specifically detects p62/SQSTM1 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunocytochemistry/Immunofluorescence, Knockout Validated.Specifications
p62/SQSTM1 | |
Polyclonal | |
Western Blot, Simple Western, Immunocytochemistry/Immunofluorescence, Knockout Validated | |
Q13501 | |
SQSTM1 | |
A synthetic peptide made to an internal region of the human p62/SQSTM1 protein (within residues 400-450). [Swiss-Prot Q13501] | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoassay | |
Biotin | |
PBS with 0.05% Sodium Azide | |
A170, EBI3-associated protein of 60 kDa, EBI3-associated protein p60, EBIAP, ORCA, OSIL, oxidative stress induced like, p60PDB3, p62, p62B, Paget disease of bone 3, phosphotyrosine independent ligand for the Lck SH2 domain p62, Phosphotyrosine-independent ligand for the Lck SH2 domain of 62 kDa, sequestosome 1, sequestosome-1, Ubiquitin-binding protein p62, ZIP3 | |
Rabbit | |
47 kDa | |
0.1 mL | |
Autophagy, Signal Transduction | |
8878 | |
Store at 4C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction