Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-p62/SQSTM1, FITC, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP142821F

Catalog No. NBP142821F


Add to cart

Description

Description

p62/SQSTM1 Polyclonal antibody specifically detects p62/SQSTM1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

p62/SQSTM1
FITC
PBS with 0.05% Sodium Azide
Affinity Purified
SQSTM1
A synthetic peptide made to an internal region of the human p62/SQSTM1 protein (within residues 350-400). [Swiss-Prot Q13501]
47 kDa
0.1 ml
Autophagy, Signal Transduction
Primary
8878
Immunocytochemistry, Immunofluorescence
Immunocytochemistry/Immunofluorescence
Q13501
A170, EBI3-associated protein of 60 kDa, EBI3-associated protein p60, EBIAP, ORCA, OSIL, oxidative stress induced like, p60PDB3, p62, p62B, Paget disease of bone 3, phosphotyrosine independent ligand for the Lck SH2 domain p62, Phosphotyrosine-independent ligand for the Lck SH2 domain of 62 kDa, sequestosome 1, sequestosome-1, Ubiquitin-binding protein p62, ZIP3
Rabbit
IgG
Immunogen affinity purified
RUO
Store at 4C in the dark.
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only