Learn More
p70 S6 Kinase Rabbit anti-Human, Mouse, Rat, Clone: 20-10C-6, MilliporeSigma™
Rabbit Monoclonal Antibody
Manufacturer:  MilliporeSigma 05781R
Catalog No. 05-781-RMI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
This Anti-p70 S6 Kinase Antibody, clone 20-10C-6 is validated for use in western blot, immunoprecipitation, IC for the detection of p70 S6 Kinase.
Specifications
Specifications
p70 S6 Kinase
20-10C-6
Purified rabbit IgG supernatant in buffer containing 0.1M Tris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide with 40% Glycerol.
Purified
KLH-conjugated linear peptide corresponding to human p70 S6 Kinase at and around the N-terminus.
Protein A purfied
−20°C for 12 months. Aliquot to avoid freeze/thaw cycles.
Monoclonal
Human, Murine, Rat
Immunocytochemistry, Immunoprecipitation, Western Blot
Unconjugated
NP_003152.1
Rabbit
IgG
100μL
Primary
RPS6KB1, p70-alpha, p70S6k, p70-S6K, p70, S6K, S6Kalpha, P70S6K, STK14A, S6K1, p70(S6K)-alpha, PS6K