Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p70 [S6K] Mouse, Unlabeled, Clone: 16, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611261

 View more versions of this product

Catalog No. BDB611261


Add to cart

Description

Description

Increased protein synthesis, an obligatory step in cell proliferation, is regulated by the phosphorylation of a number of translational components, such as ribosomal protein S6. p70 S6 Kinase (p70 [S6K]), a cytosolic kinase that is primarily responsible for S6 phosphorylation, belongs to a family of at least six S6 kinases. p70 [S6K] and p85 [S6K] are two isoforms of the same kinase. Although they are encoded by the same gene, p80 [S6K] contains a 23 amino acid N-terminal extension that serves as a nuclear localization sequence. These kinases are located in a p21 ras-independent signaling pathway and are regulated by Ser/Thr phosphorylation in response to mitogenic stimulation. Activation of p70 [S6K] requires sequential phosphorylations within the autoinhibitory domain and at Thr 389. p70 [S6K] associates with PDK-1 and PKC? to form a possible PI3-K signaling complex. The immunosuppressive agent rapamycin forms an intracellular complex with FKBP12 and FRAP (RAFT1) that inhibits phosphorylation of p70 [S6K] at Thr 389 and, in turn, prevents enzyme activation. Thus, p70 [S6K] is an important regulator of cell proliferation and a potential target of agents that modify the immune and proliferative responses. p70 [S6K] has been reported to be observable migrating within a range of 60-70 kD by western blotting.

Host Species: Mouse
Clone: 16
Isotype: IgG1
Species Reactivity: Rat
Immunogen: Rat p70 [S6K] aa. 2-121
Formula Weight [Chemical]: 60-70kDa

Immunofluorescence, Western Blotting

Specifications

Specifications

p70 [S6K]
16
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Western Blot
250μg/mL
p70 S6 Kinase
Affinity Purified
Rat p70 [S6K] aa. 2-121
150μg
Cell Biology
Primary
Human, Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.