Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
p75NTR (Neurotrophin Receptor) Mouse anti-Human, Monkey, Clone: A16/17, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05446
Description
Anti-p75NTR (Neurotrophin Receptor) Antibody, clone ME20.4 detects level of p75NTR (Neurotrophin Receptor) and has been published and validated for use in immunoprecipitation, western blot, immunohistochemistry, NEUT.Specifications
p75NTR (Neurotrophin Receptor) | |
Monoclonal | |
Unconjugated | |
NM_002507 | |
Mouse | |
Protein G chromatography | |
Primary | |
−20°C for 24 months. | |
IgG1 |
Immunohistochemistry, Immunoprecipitation, Neutralization, Western Blot | |
A16/17 | |
0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30% | |
NGFR, Gp80-LNGFR, CD271, p75(NTR), TNFRSF16 | |
Human melanoma cell line WM245. | |
100 μg | |
Human, Monkey | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction