Learn More
NGFR Rabbit anti-Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA118397
Catalog No. 01-670-649
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
PA1-18397 detects p75NTR from rat samples. PA1-18397 has been successfully used in immunohistochemistry and immunofluorescence procedures. The PA1-18397 immunogen is the extracellular domain of human p75NTR. Reconstitute with 100 μL of distilled water.
Nerve growth factor receptor contains an extracellular domain containing four 40-amino acid repeats with 6 cysteine residues at conserved positions followed by a serine/threonine-rich region, a single transmembrane domain, and a 155-amino acid cytoplasmic domain. The cysteine-rich region contains the nerve growth factor binding domain.
Specifications
Specifications
NGFR
Polyclonal
whole serum with no preservative
P07174
Low-affinity nerve growth factor receptor; NGF receptor; Gp80-LNGFR, p75 ICD, Low affinity neurotrophin receptor p75NTR, CD271 antigen, p75, p-75
Rabbit
IgG
RUO
Antibody
Polyclonal
Rat
Immunofluorescence, Immunohistochemistry
Unconjugated
Ngfr
Lyophilized
NGFR
Extracellular domain of human p75NTR.
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
24596