Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p95PKL/GIT Mouse, Unlabeled, Clone: 13, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611388

Catalog No. BDB611388


Add to cart

Description

Description

Paxillin is involved in the integration of growth factor- and adhesion-mediated signal transduction pathways at focal adhesion sites. The LD motif of paxillin has been implicated in its binding to FAK, vinculin, and paxillin-kinase linker (p95PKL). p95PKL couples paxillin to a protein complex containing p21 GTPase-activated kinase (PAK), Nck, and the guanine nucleotide exchange factor, PIX. The sequence of p95PKL, highly homologous to human GIT1 and GIT2, predicts a structure that contains an N-terminal ARF-GAP domain containing a Zn2+; finger, three ankyrin repeats, an EF hand domain, and two IQ motifs. p95PKL binds to both paxillin and PIX and colocalizes with paxillin at focal adhesion sites. Overexpression of a paxillin LD4 deletion mutant in neuroblastoma cells inhibits insulin-like growth factor-1 mediated lamellipodia formation. Microinjection of this mutant into NIH3T3 cells has been reported to decrease migration into a wound. These alterations in cell motility are thought to involve PAK-mediated regulation of the actin cytoskeleton, since the LD4 motif of paxillin is required for p95PKL-dependent recruitment of PAK/PIX complexes to paxillin-containing focal adhesions sites. This antibody has been reported to recognize GIT1 and GIT2 proteins.

Immunofluorescence, Western Blotting

Specifications

Specifications

p95PKL/GIT
13
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Western Blot
250μg/mL
Paxillin-Kinase Linker
Affinity Purified
Chicken p95PKL aa. 140-472
50μg
Cell Biology
Primary
Human, Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit