Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAAF1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524984

Catalog No. PIPA524984

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine based on sequence homology.

PAAF1 inhibits proteasome 26S assembly and proteolytic activity by impairing the association of the 19S regulatory complex with the 20S core. In case of HIV-1 infection, recruited by viral Tat to the HIV-1 promoter, where it promotes the recruitment of 19S regulatory complex through dissociation of the proteasome 26S. This presumably promotes provirus transcription efficiency.
Specifications

Specifications

PAAF1
Polyclonal
PBS with 0.09% sodium azide
Q9BRP4
PAAF, Rpn14, WDR71, WD repeat domain 71, WD repeat-containing protein 71
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
80227
Immunohistochemistry, Western Blot
Unconjugated
PAAF1
Liquid
PAAF1
KLH conjugated synthetic peptide between 109-138 amino acids from the N-terminal region of human PAAF1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit