Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PACAP Receptor Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA132888

Catalog No. PIPA132888


Add to cart

Description

Description

PA1-32888 detects PACAP receptor from human samples. PA1-32888 has been successfully used in immunohistochemistry (paraffin tissue) applications. The PA1-32888 immunogen is: Synthetic peptide, KLH conjugated.

Specifications

Specifications

PACAP Receptor
Polyclonal
Unconjugated
ADCYAP1R1
Liquid
Rabbit
IgG
25 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
117
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 0.01% sodium azide; pH 7.7
P41586
ADCYAP1R1
Synthetic peptide, KLH conjugated.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit