Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

protein kinase C and casein kinase substrate in neurons 2, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human PACSIN2 protein.

Manufacturer:  Abnova Corporation H00011252B01

Catalog No. 89-007-169


Add to cart

Description

Description

Sequence: MSVTYDDSVGVEVSSDSFWEVGNYKRTVKRIDDGHRLCSDLMNCLHERARIEKAYAQQLTEWARRWRQLVEKGPQYGTVEKAWMAFMSEAERVSELHLEVKASLMNDDFEKIKNWQKEAFHKQMMGGFKETKEAEDGFRKAQKPWAKKLKEVEAAKKAHHAACKEEKLAISREANSKADPSLNPEQLKKLQDKIEKCKQDVLKTKEKYEKSLKELDQGTPQYMENMEQVFEQCQQFEEKRLRFFREVLLEVQKHLDLSNVAGYKAIYHDLEQSIRAADAVEDLRWFRANHGPGMAMNWPQFEEWSADLNRTLSRREKKKATDGVTLTGINQTGDQSLPSKPSSTLNVPSNPAQSAQSQSSYNPFEDEDDTGSTVSEKDDTKAKNVSSYEKTQSYPTDWSDDESNNPFSSTDANGDSNPFDDDATSGTEVRVRALYDYEGQEHDELSFKAGDELTKMEDEDEQGWCKGRLDNGQVGLYPANYVEAIQ
Specifications

Specifications

protein kinase C and casein kinase substrate in neurons 2
Unconjugated
No additive
BC008037.2
SDPII
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
11252
Western Blot
Mouse polyclonal antibody raised against a full-length human PACSIN2 protein.
PACSIN2
Serum
PACSIN2
PACSIN2 (AAH08037.1, 1 a.a. ~ 486 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit