Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PACSIN3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA528708

Catalog No. PIPA528708


Add to cart

Description

Description

PA5-28708 targets PACSIN3 in WB applications and shows reactivity with Human samples. The PA5-28708 immunogen is synthetic peptide corresponding to a region within amino acids 1 and 45 of Human PACSIN3. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

This gene is a member of the protein kinase C and casein kinase substrate in neurons family. The encoded protein is involved in linking the actin cytoskeleton with vesicle formation. Alternative splicing results in multiple transcript variants.
Specifications

Specifications

PACSIN3
Polyclonal
Unconjugated
PACSIN3
Liquid
PACSIN3
Synthetic peptide corresponding to a region within amino acids 1 and 45 of Human PACSIN3
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
0.8 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q9UKS6
SDPIII, SH3 domain-containing protein 6511, protein kinase C and casein kinase substrate in neurons protein 3, syndapin III
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
29763
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit