Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PADI4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
$204.00 - $482.50
Specifications
Antigen | PADI4 |
---|---|
Concentration | 2.2 mg/mL |
Dilution | Immunohistochemistry 1:200, Immunohistochemistry-Paraffin 1:400, Western Blot (Negative), ICC/IF (Negative) |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP197309SS
|
Novus Biologicals
NBP197309SS |
0.025 mL |
Each for $204.00
|
|
|||||
NBP197309
|
Novus Biologicals
NBP197309 |
0.1 mL |
Each for $482.50
|
|
|||||
Description
PADI4 Polyclonal specifically detects PADI4 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Western Blot (Negative), ICC/IF (Negative).Specifications
PADI4 | |
Immunohistochemistry 1:200, Immunohistochemistry-Paraffin 1:400, Western Blot (Negative), ICC/IF (Negative) | |
Polyclonal | |
Rabbit | |
Lipid and Metabolism | |
PBS (pH 7.4) with 0.02% Sodium Azide | |
HL-60 PAD, PADI5PADI-H protein, PADPAD4, PDI4, PDI5EC 3.5.3.15, peptidyl arginine deiminase, type IV, peptidyl arginine deiminase, type V, Peptidylarginine deiminase IV, Protein-arginine deiminase type IV, protein-arginine deiminase type-4 | |
PADI4 | |
IgG | |
Affinity Purified |
2.2 mg/mL | |
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
RUO | |
Mouse | |
Q9UM07 | |
23569 | |
A synthetic peptide made to an internal portion of the human PAD4 protein (between residues 200-250) [UniProt Q9UM07] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title