Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-PAF1, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP219675
Description
PAF1 Polyclonal specifically detects PAF1 in Mouse samples. It is validated for Western Blot.Specifications
PAF1 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
F23149_1, FLJ11123, Paf1, RNA polymerase II associated factor, homolog (S. cerevisiae), Pancreatic differentiation protein 2, PD2hPAF1, RNA polymerase II-associated factor 1 homolog | |
Rabbit | |
60 kDa | |
0.1 mL | |
Transcription Factors and Regulators | |
54623 | |
Mouse | |
IgG |
Western Blot | |
0.37mg/mL | |
Western Blot 1:500-1:3000 | |
Q8N7H5 | |
PAF1 | |
Recombinant protein encompassing a sequence within the center region of human PAF1. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Mouse. Human (100%), Rat (100%), Bovine (100%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only