Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-PAG3, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP219678

Catalog No. NBP219678

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Specifications

Specifications

PAG3
0.77mg/mL
Western Blot 1:500-1:3000
O43150
arf-GAP with SH3 domain, ANK repeat and PH domain-containing protein 2, ArfGAP with SH3 domain, ankyrin repeat and PH domain 2, centaurin, beta 3, CENTB3, DDEF2FLJ42910, development and differentiation enhancing factor 2, Development and differentiation-enhancing factor 2, KIAA0400AMAP2, Pap-alpha, PAPPAG3, Paxillin-associated protein with ARF GAP activity 3, PYK2 C terminus-associated protein, Pyk2 C-terminus-associated protein, SHAG1
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse. Species reactivity based on sequence identity: Xenopus laevis (91%).
Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Affinity Purified
ASAP2
Recombinant protein encompassing a sequence within the N-terminus region of human DDEF2. The exact sequence is proprietary.
112 kDa
0.1 ml
Cellular Markers, Cytoskeleton Markers
Primary
8853
Human, Mouse
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only