Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAH Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524843

Catalog No. PIPA524843


Add to cart

Description

Description

This antibody is predicted to react with bovine and rat based on sequence homology.

PAH encodes the enzyme phenylalanine hydroxylase that is the rate-limiting step in phenylalanine catabolism. Deficiency of this enzyme activity results in the autosomal recessive disorder phenylketonuria.
Specifications

Specifications

PAH
Polyclonal
PBS with 0.09% sodium azide
P00439, P16331
PH, PKU, PKU1, phe-4-monooxygenase, phenylalanine 4-monooxygenase, phenylalanine-4-hydroxylase
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
18478, 5053
Western Blot
Unconjugated
PAH
Liquid
PAH
KLH conjugated synthetic peptide between 132-161 amino acids from the central region of human PAH
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit