Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERPINB2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA513501

Catalog No. PIPA513501

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PAI2 inhibits urokinase-type plasminogen activator. The monocyte derived PAI-2 is distinct from the endothelial cell-derived PAI-1.
Specifications

Specifications

SERPINB2
Polyclonal
PBS with 0.09% sodium azide
P05120, P12388
Plasminogen activator inhibitor 2; PAI-2; Placental plasminogen activator inhibitor; Monocyte Arg-serpin; Urokinase inhibitor; PLANH2; PAI2; SERPINB2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
18788, 5055
Immunohistochemistry, Western Blot
Unconjugated
SERPINB2
Liquid
SERPINB2
KLH conjugated synthetic peptide between 116-144 amino acids from the central region of human PAI2
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit