Learn More
PAK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA529224
Catalog No. PIPA529224
Add to cart
Description
Description
PA5-29225 targets TID1 in IF, IHC (P), and WB applications and shows reactivity with Human samples. The PA5-29225 immunogen is recombit fragment corresponding to a region within amino acids 172 and 438 of Human TID1.
PAK proteins are critical effectors that link RhoGTPases to cytoskeleton reorganization and nuclear signaling. PAK proteins, a family of serine/threonine p21-activating kinases, include PAK1, PAK2, PAK3 and PAK4. These proteins serve as targets for the small GTP binding proteins Cdc42 and Rac and have been implicated in a wide range of biological activities. PAK1 regulates cell motility and morphology.
Specifications
Specifications
PAK1
Polyclonal
Unconjugated
PAK1
Liquid
PAK1
Recombit fragment corresponding to a region within amino acids 167 and 436 of Human PAK1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.84 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Q13153, O88643, P35465
PAKalpha, STE20 homolog, yeast, alpha-PAK, p21/Cdc42/Rac1-activated kinase 1 (STE20 homolog, yeast), p21/Cdc42/Rac1-activated kinase 1 (yeast Ste20-related), p65-PAK, serine/threonine-protein kinase PAK 1
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
18479, 29431, 5058