Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAK1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA515115

Catalog No. PIPA515115

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine, mouse, rat, rabbit and Xenopus based on sequence homology.

PAK proteins are critical effectors that link RhoGTPases to cytoskeleton reorganization and nuclear signaling. PAK proteins, a family of serine/threonine p21-activating kinases, include PAK1, PAK2, PAK3 and PAK4. These proteins serve as targets for the small GTP binding proteins Cdc42 and Rac and have been implicated in a wide range of biological activities. PAK1 regulates cell motility and morphology.
Specifications

Specifications

PAK1
Polyclonal
PBS with 0.09% sodium azide
Q13153
Serine/threonine-protein kinase PAK 1, p21-activated kinase 1, P65-PAK, Alpha-PAK
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5058
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Unconjugated
PAK1
Liquid
PAK1
KLH conjugated synthetic peptide between 401-430 amino acids from human PAK1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit