Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAK7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA520232
Description
A suggested positive control is T24 cell lysate. PA5-20232 can be used with blocking peptide PEP-0368.
PAK7 Polyclonal antibody specifically detects PAK7 in Human samples. It is validated for Immunocytochemistry, Western Blot.Specifications
PAK7 | |
Polyclonal | |
Unconjugated | |
PAK5 | |
2900083L08Rik; 6430627N20; KIAA1264; p21 (CDKN1A)-activated kinase 7; p21 (RAC1) activated kinase 5; p21 (RAC1) activated kinase 7; p21 protein (Cdc42/Rac)-activated kinase 7; p21(CDKN1A)-activated kinase 7; p21-activated kinase 5; p21-activated kinase 7; p21CDKN1A-activated kinase 7; PAK5; PAK-5; Pak7; PAK-7; protein kinase PAK5; RP5-1119D9.3; Serine/threonine-protein kinase PAK 5; serine/threonine-protein kinase PAK 7; serine/threonine-protein kinase PAK7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57144 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q9P286 | |
PAK5 | |
A 13 amino acid peptide from near the center of human PAK5. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction