Learn More
RASSF9 Monoclonal Antibody (2F8), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA520630
Description
Peptide Sequence: MAPFGRNLLK TRHKNRSPTK DMDSEEKEIV VWVCQEEKLV CGLTKRTTSA DVIQALLEEH EATFGEKRFL LGKPSDYCII EKWRGSERVL PPLTRILKLW KAWGDEQPNM QFVLVKADAF LPVPLWRTAE AKLVQNTEKL WELSPANYMK TLPPDKQKRI VRKTFRKLAK IKQDTVSHDR DNMETLVHLI ISQDHTIHQQ VKRMKELDLE IEKCEAKFHL DRVENDGENY VQDAYLMPSF SEVEQNLDLQ YEENQTLEDL SESDGIEQLE ERLKYYRILI DKLSAEIEKE VKSVCIDINE DAEGEAASEL ESSNLESVKC DLEKSMKAGL KIHSHLSGIQ KEIKYSDSLL QMKAKEYELL AKEFNSLHIS NKDGCQLKEN RAKESEVPSS NGEIPPFTQR VFSNYTNDTD SDTGISSNHS QDSETTVGDV VLLST
RASSF9 Monoclonal antibody specifically detects RASSF9 in Human samples. It is validated for ELISA.Specifications
RASSF9 | |
Monoclonal | |
0.2-1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
O75901 | |
RASSF9 | |
RASSF9 (AAH31589, 1 a.a. ∽ 435 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA | |
2F8 | |
Unconjugated | |
RASSF9 | |
AW322107; PAM COOH-terminal interactor protein 1; Pamci; PCIP1; P-cip1; Peptidylglycine alpha-amidating monooxygenase COOH-terminal interactor; peptidylglycine alpha-amidating monooxygenase COOH-terminal interactor protein-1; Ras association (RalGDS/AF-6) domain family (N-terminal) member 9; Ras association domain family member 9; ras association domain-containing protein 9; RASSF9 | |
Mouse | |
Affinity chromatography | |
RUO | |
9182 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.