Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCN1A Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA138631

Catalog No. 01-673-754


Add to cart

Description

Description

This antibody is predicted to react with bovine, frog, and rat based on sequence homology. Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is brain tissue.

Voltage-gated sodium channels (VGSC) are present in most excitable cells. In neuronal tissue, they are responsible for generating and propagating action potentials. Brain VGSCs are heteromers of alphabeta1beta2 subunits. Of these, the alpha subunit forms the channel pore. This antibody recognizes the alpha subunit of type I voltage-gated sodium channel.
Specifications

Specifications

SCN1A
Polyclonal
PBS with 1% BSA and 0.1% sodium azide
P35498, Q9NY46, Q9UI33, Q9UQD0, Q9Y5Y9, A2APX8, P35499, Q14524, Q15858, Q99250
EIEE6, FEB3, FEB3A, FHM3, GEFSP2, HBSCI, NAC1, Nav1.1, SCN1, SMEI, sodium channel protein brain I subunit alpha, sodium channel protein type 1 subunit alpha, sodium channel protein type I subunit alpha, sodium channel protein, brain I alpha subunit, sodium channel voltage gated type 1 alpha subunit, sodium channel, voltage-ga
Rabbit
IgG
1 mL
-20° C, Avoid Freeze/Thaw Cycles
Primary
6329, 6331, 6335, 6323, 6328, 6336, 11280, 20265, 6326, 6334
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
SCN1A
Liquid
SCN10A, SCN11A, SCN1A, SCN2A, SCN3A, SCN4A, SCN5A, SCN8A, SCN9A
Synthetic peptide from C-terminus of the alpha subunit of rat type 1 voltage-gated sodium channel.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit