Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

pan Synuclein Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA138705

Catalog No. 01-674-694


Add to cart

Description

Description

This antibody is predicted to react with rat samples based on sequence homology. Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is brain tissue.

Synucleins are small, soluble proteins expressed primarily in neural tissue and in certain tumors. The family includes three known proteins: alpha-synuclein, beta-synuclein, and gamma-synuclein. The alpha- and beta-synuclein proteins are found primarily in brain tissue, where they are seen mainly in presynaptic terminals. The gamma-synuclein protein is found primarily in the peripheral nervous system and retina, but its expression in breast tumors makes it a candidate as a marker for tumor progression.
Specifications

Specifications

pan Synuclein
Polyclonal
PBS with 1% BSA and 0.1% sodium azide; pH 7.6
O76070, P37840, Q16143
PD1, NACP, PARK1, PARK4, SR, BCSG1
Rabbit
IgG
1 mL
-20° C, Avoid Freeze/Thaw Cycles
Primary
6620, 6622, 6623
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
SNCA
Liquid
SNCA, SNCB, SNCG
Synthetic peptide near N-terminus of human alpha, beta, gamma synuclein protein
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit