Learn More
MilliporeSigma™ anti-PAN TORC (1-42), Polyclonal
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma™ ST109850UL
Catalog No. ST109850UL
Add to cart
Description
Description
Specifically detects PAN TORC (1-42) Clone: in Human, Mouse, Rat samples, and it is validated for Immunoprecipitation
TORCs (Transducers Of Regulated cAMP Response Element-Binding (CREB) protein) are members of a conserved family of co-activators that enhance CRE-dependent transcription by a phosphorylation-independent interaction with the bZIP DNA binding/dimerization domain of CREB. Studies suggest that TORC phosphorylation and nuclear/cytoplasmic shuttling play an important role in the regulation of gluconeogenesis by cAMP.
Specifications
Specifications
PAN TORC (1-42)
Unconjugated
Undiluted Serum
a recombinant protein consisting of amino acids 1-42 of human TORC1 fused to GST
50μL
2°C to 8°C
Polyclonal
Human, Murine, Rat
Immunoprecipitation
Undiluted serum.
Rabbit
IgG
RUO
Primary
Recognizes the ∽79kDa TORC1, TORC2, and TORC3 proteins in 293T cells over-expressing TORC1, TORC2, or TORC3. Recognizes endogenous TORC1 and TORC2.