Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAN3 Rabbit anti-Human, Polyclonal, Invitrogen
Rabbit Polyclonal Antibody
Manufacturer: Invitrogen PA524880
Description
This antibody is predicted to react with mouse based on sequence homology.
PAN3 is functions in cytoplasmic mRNA decay. It is as part of the Pan nuclease complex, recruits polyadenylate-binding protein which in turn stimulates PAN2 nuclease activity.Specifications
PAN3 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q58A45 | |
PAB-dependent poly(A)-specific ribonuclease subunit 3, PABP-dependent poly(A) nuclease 3, PABP1-dependent poly A-specific ribonuclease subunit PAN3, hPan3 | |
Rabbit | |
IgG | |
400 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
255967 |
Flow Cytometry, Immunohistochemistry, Western Blot | |
Unconjugated | |
PAN3 | |
Liquid | |
PAN3 | |
KLH conjugated synthetic peptide between 771-799 amino acids from the C-terminal region of human PAN3 | |
Antigen affinity chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title