Learn More
Chromogranin A Rabbit anti-Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA136153
Catalog No. 01-672-106
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Reconstitute with 50-100 µL of distilled water, and dilute further in 0.1M PBS with 1% BSA and 0.1% sodium azide.
Chromogranin A (CHGA) is a member of the chromogranin/secretogranin family of neuroendocrine secretory proteins. It is found in secretory vesicles of neurons and endocine cells. CHGA is a precursor to three biologically active peptides: vasostatin, pancreastatin, and parastatin. These peptides act as autocine or paracrine negative modulators of the neuroendocrine system. CHGA is an excellent marker for carcinoid tumors, pheo-chromocytomas, paragangliomas, and other neuro-endocrine tumors. Coexpression of chromogranin A and neuron specific enolase (NSE) is common in neuroendocrine neoplasms.
Specifications
Specifications
Chromogranin A
Polyclonal
whole serum with 0.1% sodium azide
P10354
MGC105435
Rabbit
IgG
RUO
Antibody
Polyclonal
Rat
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Unconjugated
Chga
Lyophilized
CHGA
Synthetic rat chromogranin A (residues 306-314).
50 μL
4° C
Primary
24258