Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

panMunc13 Mouse, Unlabeled, Clone: 32, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610999

 View more versions of this product

Catalog No. BDB610999


Add to cart

Description

Description

Release of neurotransmitters at the synaptic junction is a primary mechanism of neuronal communication and is regulated by the synaptic vesicle (SV) cycle. Several components of the SV cycle were initially characterized in C. elegans. One of those, unc-13, is involved in normal presynaptic function. Mammalian homologues of unc-13 are Munc13-1, 13-2, and 13-3. These neuron-specific proteins lack N-terminal homology, but are highly conserved in their C-terminal regions. Here they contain a phorbal ester binding C1-domain and two C2-domains with homology to the PKC Ca2+;/phospholipid-binding domain. The most abundant isoform, Munc 13-1, is a presynaptic receptor with high affinities for phorbol ester and DAG. In response to phorbol ester, Munc13-1 associates with the plasma membrane and acts as a stimulation dependent enhancer of neurotransmitter release. In addition, Munc13-1 interacts with components of the SV cycle (syntaxin, SNAP25, synaptobrevin, Doc2) and with a brain specific isoform of β-spectrin, a protein that interacts with the actin cytoskeleton. Thus, it is thought that Munc13 proteins function in signaling pathways that regulate the neuronal exocytic machinery.

Immunofluorescence, Western Blotting

Specifications

Specifications

panMunc13
32
Unconjugated
Affinity Purified
Rat Munc13-1 aa. 621-834
150μg
Cell Biology
Primary
Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.