Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAP-Goat Goat anti-Goat, Polyclonal, MilliporeSigma™

Goat Polyclonal Antibody

Manufacturer:  MilliporeSigma PAP10250UL

Catalog No. PAP10250UL


Add to cart

Description

Description

PAP-Goat is an antibody targeting the PAP-Goat protein, validated for use in immunohistochemistry.
Specifications

Specifications

PAP-Goat
20mg/mL
Lyophilized. Buffer = 0.01M Sodium phosphate, 0.25M NaCl containing 0.01% thimerosal, pH 7.6.

RECONSTITUTION: Reconstitute to proper volume with sterile distilled water. Let stand 1-2 hours at room temperature.
Goat
Lyophilized: 2-8°C for up to 12 months. Reconstituted: −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Avoid repeated freeze/thaw cycles.
Polyclonal
Immunohistochemistry
Unconjugated
Purified
250μL
Primary
Goat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only