Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3'-phosphoadenosine 5'-phosphosulfate synthase 2, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human PAPSS2 protein.

Manufacturer:  Abnova Corporation H00009060B01

Catalog No. 89-006-698


Add to cart

Description

Description

Sulfation is a common modification of endogenous (lipids, proteins, and carbohydrates) and exogenous (xenobiotics and drugs) compounds. In mammals, the sulfate source is 3'-phosphoadenosine 5'-phosphosulfate (PAPS), created from ATP and inorganic sulfate. Two different tissue isoforms encoded by different genes synthesize PAPS. This gene encodes one of the two PAPS synthetases. Defects in this gene cause the Pakistani type of spondyloepimetaphyseal dysplasia. Two alternatively spliced transcript variants that encode different isoforms have been described for this gene. [provided by RefSeq

Sequence: MSGIKKQKTENQQKSTNVVYQAHHVSRNKRGQVVGTRGGFRGCTVWLTGLSGAGKTTISFALEEYLVSHAIPCYSLDGDNVRHGLNRNLGFSPGDREENIRRIAEVAKLFADAGLVCITSFISPFAKDRENARKIHESAGLPFFEIFVDAPLNICESRDVKGLYKRARAGEIKGFTGIDSDYEKPETPERVLKTNLSTVSDCVHQVVELLQEQNIVPYTIIKDIHELFVPENKLDHVRAEAETLPSLSITKLDLQWVQVLSEGWATPLKGFMREKEYLQVMHFDTLLDDGVINMSIPIVLPVSAEDKTRLEGCSKFVLAHGGRRVAILRDAEFYEHRKEERCSRVWGTTCTKHPHIKMVMESGDWLVGGDLQVLEKIRWNDGLDQYRLTPLELKQKCKEMNADAVFAFQLRNPVHNGHALLMQDTRRRLLERGYKHPVLLLHPLGGWTKDDDVPLDWRMKQHAAVLEEGVLDPKSTIVAIFPSPMLYAGPTEVQWHCRSRMIAGANFYIVGRDPAGMPHPETKKDLYEPTHGGKVLSMAPGLTSVEIIPFRVAAYNKAKKAMDFYDPARHNEFDFISGTRMRKLAREGENPPDGFMAPKAWKVLTDYYRSLEKN
Specifications

Specifications

3'-phosphoadenosine 5'-phosphosulfate synthase 2
Unconjugated
No additive
NM_004670.3
ATPSK2|SK2
Mouse
50 uL
Stem Cell Biology
Y
Polyclonal
Human
Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Mouse polyclonal antibody raised against a full-length human PAPSS2 protein.
PAPSS2
Serum
PAPSS2
PAPSS2 (NP_004661.2, 1 a.a. ~ 614 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
9060
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit