Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Parainfluenza Virus Type 1 Goat anti-Virus, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA17236

Catalog No. PA17236


Add to cart

Description

Description

PA1-7236 detects the parainfluenza virus, type 1. This antibody does not cross react with parainfluenza types 2 and 3. PA1-7236 has been successfully used in immunofluorescence procedures. The PA1-7236 immunogen is Cantell strain.

Specifications

Specifications

Parainfluenza Virus Type 1
Polyclonal
Unconjugated
Liquid
Goat
IgG
1 mL
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Primary
Virus
Immunofluorescence
4-5 mg/mL
PBS with 0.1% sodium azide; pH 7.2
hPIV-1; PIV-1; Parainfluenza Virus Type 1
Cantell strain.
Purified
RUO
Antibody
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit