Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Parainfluenza Virus Type 2/3 Goat anti-Virus, FITC, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA173039

Catalog No. PIPA173039


Add to cart

Description

Description

The PA1-73039 antibody reacts with Parainfluenza types 2 & 3. PA1-73039 has been successfully used in Immunofluorescence applications. The PA1-73039 antibody was raised against Parainfluenza type 3 isolated from human serum.

Specifications

Specifications

Parainfluenza Virus Type 2/3
Polyclonal
FITC
Liquid
Goat
IgG
1 mL
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Primary
Virus
Immunofluorescence
4-5 mg/mL
PBS with 10mg/mL BSA and 0.1% sodium azide
hPIV-2; PIV-2; Parainfluenza Virus Type 2; PIV-3; Parainfluenza Virus Type 3
Parainfluenza type 3 isolated from human serum.
Purified
RUO
Antibody
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit