Learn More
Please call 1-800-766-7000 to place your order or try our site again later.
par-6 partitioning defective 6 homolog alpha (C. elegans) (B01), Mouse anti-Human, Polyclonal Antibody, Abnova
Mouse polyclonal antibody raised against a full-length human PARD6A protein.
Manufacturer: Abnova Corporation H00050855B01
Description
This gene is a member of the PAR6 family and encodes a protein with a PSD95/Discs-large/ZO1 (PDZ) domain and a semi-Cdc42/Rac interactive binding (CRIB) domain. This cell membrane protein is involved in asymmetrical cell division and cell polarization processes as a member of a multi-protein complex. The protein also has a role in the epithelial-to-mesenchymal transition (EMT) that characterizes the invasive phenotype associated with metastatic carcinomas. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq
Sequence: MARPQRTPARSPDSIVEVKSKFDAEFRRFALPRASVSGFQEFSRLLRAVHQIPGLDVLLGYTDAHGDLLPLTNDDSLHRALASGPPPLRLLVQKREADSSGLAFASNSLQRRKKGLLLRPVAPLRTRPPLLISLPQDFRQVSSVIDVDLLPETHRRVRLHKHGSDRPLGFYIRDGMSVRVAPQGLERVPGIFISRLVRGGLAESTGLLAVSDEILEVNGIEVAGKTLDQVTDMMVANSHNLIVTVKPANQRNNVVRGASGRLTGPPSAGPGPAEPDSDDDSSDLVIENRQPPSSNGLSQGPPCWDLHPGCRHPGTRSSLPSLDDQEQASSGWGSRIRGDGSGFSLSpecifications
par-6 partitioning defective 6 homolog alpha (C. elegans) | |
Unconjugated | |
No additive | |
NM_001037281 | |
PAR-6A|PAR6|PAR6C|PAR6alpha|TAX40|TIP-40 | |
Mouse | |
50 uL | |
Cell Cycle | |
Y | |
Polyclonal | |
Human |
Western Blot | |
Mouse polyclonal antibody raised against a full-length human PARD6A protein. | |
PARD6A | |
Serum | |
PARD6A | |
PARD6A (NP_001032358, 1 a.a. ~ 345 a.a) full-length human protein. | |
RUO | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Primary | |
50855 |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.