Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PARG1, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA127663

Catalog No. PIPA127663

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PA1-27663 detects PARG1 from human samples.PA1-27663 may be successfully used in Western blot applications.The immunogen is a synthetic peptide corresponding to human PTPL1-associated RhoGAP.
Specifications

Specifications

PARG1
Polyclonal
PBS with no preservative
Q52LW3
PARG1, RP11-255E17.1
Chicken
IgY
50μg
Store at -20°C
Primary
9411
Western Blot
Unconjugated
ARHGAP29
Purified
ARHGAP29
Synthetic peptide corresponding to human PTPL1-associated RhoGAP.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit