Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PARK7, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against a full-length human PARK7 protein.

Manufacturer:  Abnova Corporation H00011315D01P

Catalog No. 89-002-985


Add to cart

Description

Description

The product of this gene belongs to the peptidase C56 family of proteins. It acts as a positive regulator of androgen receptor-dependent transcription. It may also function as a redox-sensitive chaperone, as a sensor for oxidative stress, and it apparently protects neurons against oxidative stress and cell death. Defects in this gene are the cause of autosomal recessive early-onset Parkinson disease 7. Two transcript variants encoding the same protein have been identified for this gene. [provided by RefSeq

Sequence: MASKRALVILAKGAEEMETVIPVDVMRRAGIKVTVAGLAGKDPVQCSRDVVICPDASLEDAKKEGPYDVVVLPGGNLGAQNLSESAAVKEILKEQENRKGLIAAICAGPTALLAHEIGFGSKVTTHPLAKDKMMNGGHYTYSENRVEKDGLILTSRGPGTSFEFALAIVEALNGKEVAAQVKAPLVLKD
Specifications

Specifications

PARK7
Unconjugated
In 1X PBS, pH 7.4
NM_007262.3
DJ-1|DJ1|FLJ27376|FLJ34360|FLJ92274
Rabbit
100 ug
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
Immunofluorescence, Western Blot
Rabbit polyclonal antibody raised against a full-length human PARK7 protein.
PARK7
Purified
PARK7
PARK7 (NP_009193.2, 1 a.a. ~ 189 a.a) full-length human protein.
RUO
Primary
11315
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit