Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DJ-1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA137560

Catalog No. 01-673-367


Add to cart

Description

Description

This antibody is predicted to react with bovine, chicken, canine, mouse, porcine and rat based on sequence homology. Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10-20 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is normal breast tissue.

DJ-1 protein has 189 amino acids and was cloned from a HeLa cell cDNA library as a novel oncogene which weakly transformed NIH3T3 cells. The expression of DJ-1 is induced by growth signals and is translocated from the cytoplasm to the nucleus during the S phase, although no nuclear localization signals are present. It has been suggested that DJ-1 is a mitogen-dependent oncogene product involved in Ras-related signal transduction pathway.
Specifications

Specifications

DJ-1
Polyclonal
PBS with 1% BSA and 0.1% sodium azide
Q99497
DJ-1, DJ1, Parkinson disease autosomal recessive, early onset 7, oncogene DJ1, protein DJ-1
Rabbit
IgG
1 mL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
11315
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PARK7
Liquid
PARK7
Synthetic peptide derived from C-terminus of human DJ-1 protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit