Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DJ-1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA14369

Catalog No. PA14369


Add to cart

Description

Description

PA1-4369 detects PARK7 from human and mouse samples. PA1-4369 has been successfully used in Western blot and immunohistochemistry procedures. The PA1-4369 immunizing peptide corresponds to amino acid residues 167-189 from human PARK7.

Four chromosomal loci (PARK2, PARK6, PARK7, and PARK9) associated with autosomal recessive, early onset parkinsonism are known. Positional cloning within the refined PARK7 critical region recently identified mutations in the DJ-1 gene in PARK7-linked families. Mutations in the PARK7/DJ-1 gene cause autosomal-recessive Parkinsons disease.
Specifications

Specifications

DJ-1
Polyclonal
Unconjugated
PARK7
Liquid
PARK7
Synthetic peptide corresponding to residues A(167) I V E A L N G K E V A A Q V K A P L V L K D(189) of human PARK7.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry, Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q99497, Q99LX0
DJ-1, Oncogene DJ1
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
11315, 57320
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit