Learn More
Parkin Mouse anti-Human, Mouse, Clone: PRK8, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05882
Catalog No. 05-882-MI
Add to cart
Description
Description
Anti-Parkin Antibody, clone PRK8 detects level of Parkin and has been published and validated for use in immunoprecipitation, western blot, immunocytochemistry, immunohistochemistry.
Specifications
Specifications
Parkin
PRK8
NM_004562
PARK2, PDJ, Parkin, AR-JP, PRKN, LPRS2, parkin
Full length recombinant human Parkin. Epitope has been mapped to the second ring domain (aa 399-465)
100μL
Primary
Human, Murine
Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Unconjugated
Ascites
Murine
Ascites
−20°C for up to 24 months.
Monoclonal
Documents
Promotions
Promotions
  • MilliporeSigma
    Buy two qualifying MilliporeSigma antibodies and get a third of equal or lesser value at no additional cost.
    01-31-2021