Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-PARP-, Clone: 4G8, ™

Supplier:  MilliporeSigma™ MABE18/DEL

Encompass

Catalog No. 50-175-906

Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative.


Description

Description

Poly(ADP-ribose) polymerase (PARP) is a highly conserved nuclear enzyme present in higher eukaryotes. This gene is a member of a family of enzymes that synthesize and transfer ADP-ribose polymers to protein to modify their functional properties. A key role of PARP in the cell is mediating the repair of single-strand DNA nicks. PARP-2 forms homodimers and hetero dimers with PARP-1 in the base excision repair (BER) pathway. To mediate repair PARP interacts with DNA and recruits DNA repair protein XRCC1, DNA ligase III, DNA polymerase beta and a kinase to the site of damage. As a marker for apoptosis, it has been shown that PARP cleavage mediated by caspase-3 activity occurs at the onset of apoptosis, and inhibition of PARP cleavage attenuates apoptosis in vitro.

Host Species: Mouse
Clone: 4G8
Isotype: IgG1, κ
Cross Reactivity: Human, Mouse
Immunogen: Recombinant protein corresponding to mouse PARP-2.

ELISA, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

PARP-
4G8
Mouse
100 μg
Human, Mouse
IgG1 κ
Monoclonal
Unconjugated
Recombinant protein corresponding to mouse PARP-2.
Primary
Purified
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit