Learn More
PARP16 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA525891
Catalog No. PIPA525891
Add to cart
Description
Description
The function of this protein has not been specifically defined.
Specifications
Specifications
PARP16
Polyclonal
Unconjugated
PARP16
Liquid
PARP16
KLH conjugated synthetic peptide between 215-244 amino acids from the C-terminal region of human PARP16
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Flow Cytometry, Immunohistochemistry, Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q8N5Y8
C15orf30, pART15, PARP-16, poly [ADP-ribose] polymerase 16
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
54956