Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
parvin, gamma, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human PARVG protein.
Manufacturer: Abnova Corporation H00064098B01
Description
Members of the parvin family, including PARVG, are actin-binding proteins associated with focal contacts.[supplied by OMIM
Sequence: MEPEFLYDLLQLPKGVEPPAEEELSKGGKKKYLPPTSRKDPKFEELQKVLMEWINATLLPEHIVVRSLEEDMFDGLILHHLFQRLAALKLEAEDIALTATSQKHKLTVVLEAVNRSLQLEEWQAKWSVESIFNKDLLSTLHLLVALAKRFQPDLSLPTNVQVEVITIESTKSGLKSEKLVEQLTEYSTDKDEPPKDVFDELFKLAPEKVNAVKEAIVNFVNQKLDRLGLSVQNLDTQFADGVILLLLIGQLEGFFLHLKEFYLTPNSPAEMLHNVTLALELLKDEGLLSCPVSPEDIVNKDAKSTLRVLYGLFCKHTQKAHRDRTPHGAPNSpecifications
parvin, gamma | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human PARVG protein. | |
PARVG | |
PARVG | |
PARVG (NP_071424.1, 1 a.a. ∼ 331 a.a) full-length human protein. | |
RUO | |
Primary | |
Human |
Western Blot | |
Unconjugated | |
No additive | |
NM_022141.4 | |
Mouse | |
50 μL | |
Yes | |
64098 | |
Serum |
Safety and Handling
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title