Learn More
PASK Mouse anti-Human, Non-human primate, Rat, Clone: mAB6, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA1700
Catalog No. 01-670-127
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
MA1-700 contains 100 μg of Protein A/G purified, in vitro produced IgG2a (1 mg/mL) in PBS with 1 mg/mL BSA and 0.05% sodium azide. MA1-700 detects PASK from human, non-human primate and rat samples. MA1-700 has been successfully used in Western blot and CC/IF procedures. The MA1-700 immunogen is Recombinant, GST tagged, PAS fusion protein expressed in E. coli This monoclonal antibody was expressed by hybridoma cells grown in vitro in ABR’s dialysis-based bioreactor system. All media, reagents, and other variables that affect the system are continuously monitored and controlled, ensuring a superior degree of precision and repeatability compared to an in vivo ascites production. Additionally, the serum free medium is further protein purified for improved antibody performance.
PASKIN (PAS-Kinase) is thought to regulate protein synthesis in cells. It is a conserved gene product found in yeast as well as mammals. PASKIN contains two domains (PAS A and PAS B) as well as a serine/threonine kinase domain related to AMP kinases. PASKIN activity leads to decreased carbohydrate storage, as well as increased protein synthesis. PASKIN-dependent phosphorylation inhibits the activity of mammalian glycogen synthase.
Specifications
Specifications
PASK
Monoclonal
1 mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide
Q96RG2
PAS-Kinase
Mouse
IgG2a
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
23178, 301617
Immunocytochemistry, Immunofluorescence, Western Blot
mAB6
Unconjugated
PASK
Liquid
PASK
Recombit, GST tagged, PAS fusion protein expressed in E. coli.
Protein A
RUO
Antibody
Monoclonal
Human, Non-human Primate, Rat