Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAT1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA514143

Catalog No. PIPA514143

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with mouse and rat based on sequence homology.

APPBP2 interacts with microtubules and is functionally associated with beta-amyloid precursor protein transport and/or processing. The beta-amyloid precursor protein is a cell surface protein with signal-transducing properties, and it is thought to play a role in the pathogenesis of Alzheimer ft.s disease. This protein has been found to be highly expressed in breast cancer.
Specifications

Specifications

PAT1
Polyclonal
PBS with 0.09% sodium azide
Q92624
Amyloid protein-binding protein 2, Amyloid beta precursor protein-binding protein 2, APP-BP2, Protein interacting with APP tail 1, APPBP2, KIAA0228, PAT1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10513
Western Blot
Unconjugated
APPBP2
Liquid
APPBP2
KLH conjugated synthetic peptide between 242-271 amino acids from the central region of human PAT1(APPBP2)
Protein G
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit