Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PATL1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526797

Catalog No. PIPA526797

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with mouse and rat based on sequence homology.

Specifications

Specifications

PATL1
Polyclonal
PBS with 0.09% sodium azide
Q86TB9
Pat1b, hPat1b, PAT1-like protein 1, protein PAT1 homolog 1, protein PAT1 homolog b
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
219988
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
PATL1
Liquid
PATL1
KLH conjugated synthetic peptide between 489-518 amino acids from the C-terminal region of human PATL1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit