Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZG16B Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524448

Catalog No. PIPA524448

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

ZG16B
Polyclonal
PBS with 0.09% sodium azide
Q96DA0
EECP, HRPE773, JCLN2, PAUF, PRO1567, endocrine and exocrine protein, jacalin-like lectin domain containing 2, pancreatic adenocarcinoma upregulated factor, zymogen granule protein 16 homolog B
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
124220
Western Blot
Unconjugated
ZG16B
Liquid
ZG16B
KLH conjugated synthetic peptide between 109-137 amino acids from the central region of human PAUF
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit