Learn More
Pax5 Rat anti-Mouse, Human, Clone: 1H9, MilliporeSigma™
Rat Monoclonal Antibody
Supplier:  MilliporeSigma 051573
Catalog No. 05-157-3MI
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
Detect PAX5 using this rat monoclonal antibody, Anti-Pax5 Antibody, clone 1H9 validated for use in western blotting and immunohistochemistry.
Specifications
Specifications
Pax5
Monoclonal
Unconjugated
NP_032808
Rat
Protein G purified
Primary
2-8°C for one year
IgG2a κ
Immunohistochemistry, Western Blot
1H9
Purified rat monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Pax5, Pax-5
GST-tagged recombinant protein corresponding to mouse Pax5.
100 μg
Human, Mouse
Purified
Product Suggestions
Product Suggestions