Learn More
PAX6 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA525970
Catalog No. PIPA525970
Add to cart
Description
Description
PAX6 encodes paired box gene 6, one of many human homologs of the Drosophila melanogaster gene prd. In addition to the hallmark feature of this gene family, a conserved paired box domain, the encoded protein also contains a homeo box domain. Both domains are known to bind DNA, and function as regulators of gene transcription. This gene is expressed in the developing nervous system, and in developing eyes. Mutations in this gene are known to cause ocular disorders such as aniridia and Peter ft.s anomaly.
Specifications
Specifications
PAX6
Polyclonal
Unconjugated
PAX6
Liquid
PAX6
KLH conjugated synthetic peptide between 352-380 amino acids from human PAX6
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
P26367
AN, AN2, D11S812E, MGDA, WAGR, aniridia type II protein, oculorhombin, paired box homeotic gene-6, paired box protein Pax-6
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5080