Learn More
PAX8 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA527461
Catalog No. PIPA527461
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
PA5-27462 targets Annexin VI in IHC (P) and WB applications and shows reactivity with Human and mouse samples. The PA5-27462 immunogen is recombit fragment corresponding to a region within amino acids 475 and 673 of Human Annexin VI.
This gene encodes a member of the paired box (PAX) family of transcription factors. Members of this gene family typically encode proteins that contain a paired box domain, an octapeptide, and a paired-type homeodomain. This nuclear protein is involved in thyroid follicular cell development and expression of thyroid-specific genes. Mutations in this gene have been associated with thyroid dysgenesis, thyroid follicular carcinomas and atypical follicular thyroid adenomas. Alternateively spliced transcript variants encoding different isoforms have been described.
Specifications
Specifications
PAX8
Polyclonal
Unconjugated
PAX8
Liquid
PAX8
Recombit protein fragment corresponding to a region within amino acids 155 and 366 of Human PAX8C
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q06710
paired box protein Pax-8, paired domain gene 8
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
7849