Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Paxillin Mouse, TRITC, Clone: 349, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610055

Catalog No. BDB610055


Add to cart

Description

Description

A number of cytoskeletal proteins are tyrosine phosphorylated in Rous sarcoma virus-transformed chick embryo fibroblasts. One of these is the 68 kDa paxillin protein. Paxillin is a cytoskeletal component that localizes to the focal adhesions at the ends of actin stress fibers. It is also present in the focal adhesions of Madin-Darby canine kidney epithelial cells, but is absent from the cell adherens junctions of these cells. Paxillin purified from chicken gizzard migrates as a diffuse band on SDS-PAGE with molecular weight of 65-70 kDa. It binds to the rod domain of vinculin, another focal adhesion protein. It is thought that phosphorylation of paxillin may have a role in that disassembly of focal adhesions and stress fibers during transformation.
Specifications

Specifications

Paxillin
349
TRITC
Affinity Purified
Chicken Paxillin aa. 1 to 557
50μg
Cell Biology
Primary
Human, Murine
Immunofluorescence
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit