Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-PBEF/Visfatin/NAMPT, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP223667

 View more versions of this product

Catalog No. NBP223667


Add to cart

Description

Description

PBEF/Visfatin/NAMPT Polyclonal specifically detects PBEF/Visfatin/NAMPT in Human, Mouse, Rat, Chicken, Primate samples. It is validated for Western Blot.
Specifications

Specifications

PBEF/Visfatin/NAMPT
Unconjugated
Crude whole rabbit sera with 50% Glycerol with 0.025% Sodium Azide
Antisera
NAMPT
A portion of amino acid 30-80 of human NAMPT/Visfatin was used as the immunogen.
Unpurified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
The amino acid sequence used as immunogen is 100% homologous in human, monkey, chimpanzee and chicken, and 94% homologous in horse, cow, mouse and rat and 83% homologous in zebrafish. The NAMPT protein has multiple variants of varying molecular weights including ∽55.5kDa (alternative variant a), 41.9kDa (alternative variant b), 35.6 kDa alternative variant c), 28.5kDa (alternative variant d), and 26.8 (alternative variant e). NBP2-23795 detects an ∽38 kDa band in western blot in the cells and tissues screened. This band may correspond to NAMPT, alternative variants b or c, or another NAMPT form.
Western Blot
Western Blot 1:1000 - 1:5000
P43490
EC 2.4.2.12, MGC117256, NAmPRTase, Nampt, nicotinamide phosphoribosyltransferase, PBEF1110035O14Rik, PBEF1DKFZp666B131, Pre-B cell-enhancing factor, pre-B-cell colony enhancing factor 1, Pre-B-cell colony-enhancing factor 1, VF, VISFATIN
Rabbit
IgG
0.1 ml
Adaptive Immunity, Cell Cycle and Replication, Immunology, Innate Immunity
Primary
10135
Human, Mouse, Rat, Chicken, Primate
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only