Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PBEF/Visfatin/NAMPT Rabbit anti-Human, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$119.60 - $261.50

Specifications

Host Species Rabbit
Isotype IgG
Immunogen Amino acid 395-412 FKDPVADPNKRSKKGRLS of human NAMPT protein were used as the immunogen.
Antigen PBEF/Visfatin/NAMPT
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP223795SS
View Documents
Novus Biologicals
NBP223795SS
0.025 ml
Each for $119.60
Add to cart
 
NBP223795
View Documents
Novus Biologicals
NBP223795
0.1 ml
Each for $261.50
Add to cart
 
Description

Description

PBEF/Visfatin/NAMPT Polyclonal antibody specifically detects PBEF/Visfatin/NAMPT in Human, Rat samples. It is validated for Western Blot.
Specifications

Specifications

Rabbit
Amino acid 395-412 FKDPVADPNKRSKKGRLS of human NAMPT protein were used as the immunogen.
Polyclonal
Primary
Western Blot
P43490
Crude whole antisera with 0.02% Sodium Azide
IgG
PBEF/Visfatin/NAMPT
Antisera
RUO
The amino acid sequence used as immunogen is 100% homologous in human, monkey, horse, cow, rat and dog, 81% homologous in pufferfish and 71% homologous in zebrafish.
10135
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit